Assay Details
| Gene Symbol: GC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101679.4 | NP_173257.2 | 2 - 3 | 1267 | 68 | |
| GenBank mRNA | EU249330.1 | - | 2 - 3 | 1091 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GC2| Gene Symbol: GC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101679.4 | NP_173257.2 | 2 - 3 | 1267 | 68 | |
| GenBank mRNA | EU249330.1 | - | 2 - 3 | 1091 | 68 | |
Set Membership: |
|