Assay Details
| Gene Symbol: GC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101679.4 | NP_173257.2 | 7 - 8 | 1640 | 126 | |
| GenBank mRNA | EU249330.1 | - | 7 - 8 | 1464 | 126 | |
More Information
Set Membership: |
|

GC2
Genomic Map