Assay Details
| Gene Symbol: GC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101679.4 | NP_173257.2 | 9 - 10 | 1817 | 98 | |
| GenBank mRNA | EU249330.1 | - | 9 - 10 | 1641 | 98 | |
More Information
Set Membership: |
|

GC2
Genomic Map