Assay Details
| Gene Symbol: EBS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101684.4 | NP_564049.1 | 2 - 3 | 1154 | 129 | |
| GenBank mRNA | AB189471.1 | - | 2 - 3 | 1077 | 129 | |
More Information
Set Membership: |
|

EBS5
Genomic Map