Assay Details
| Gene Symbol: HIK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101695.5 | NP_173273.2 | 5 - 6 | 821 | 90 | |
| GenBank mRNA | AB081599.1 | - | 4 - 5 | 554 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIK| Gene Symbol: HIK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101695.5 | NP_173273.2 | 5 - 6 | 821 | 90 | |
| GenBank mRNA | AB081599.1 | - | 4 - 5 | 554 | 90 | |
Set Membership: |
|