Assay Details
| Gene Symbol: HIK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101695.5 | NP_173273.2 | 7 - 8 | 961 | 65 | |
| GenBank mRNA | AB081599.1 | - | 6 - 7 | 694 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HIK| Gene Symbol: HIK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101695.5 | NP_173273.2 | 7 - 8 | 961 | 65 | |
| GenBank mRNA | AB081599.1 | - | 6 - 7 | 694 | 65 | |
Set Membership: |
|