Assay Details
| Gene Symbol: BEE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332342.1 | NP_001321164.1 | 1 - 2 | 422 | 130 | |
| NM_101698.3 | NP_173276.2 | 1 - 2 | 422 | 130 | ||
| GenBank mRNA | AK117269.1 | - | 1 - 2 | 422 | 130 | |
| AY138253.1 | - | 1 - 2 | 393 | 130 | ||
| BT005268.1 | - | 1 - 2 | 356 | 130 | ||
More Information
Set Membership: |
|

BEE1
Genomic Map