Assay Details
| Gene Symbol: ARP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101702.4 | NP_564051.1 | 12 - 13 | 986 | 73 | |
| GenBank mRNA | AK228098.1 | - | 12 - 13 | 925 | 73 | |
| BT006474.1 | - | 11 - 12 | 835 | 73 | ||
More Information
Set Membership: |
|

ARP4
Genomic Map