Assay Details
| Gene Symbol: ARP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101702.4 | NP_564051.1 | 3 - 4 | 419 | 93 | |
| GenBank mRNA | AK228098.1 | - | 3 - 4 | 358 | 93 | |
| BT006474.1 | - | 2 - 3 | 268 | 93 | ||
More Information
Set Membership: |
|

ARP4
Genomic Map