Assay Details
| Gene Symbol: ARP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101702.4 | NP_564051.1 | 8 - 9 | 736 | 73 | |
| GenBank mRNA | AK228098.1 | - | 8 - 9 | 675 | 73 | |
| BT006474.1 | - | 7 - 8 | 585 | 73 | ||
More Information
Set Membership: |
|

ARP4
Genomic Map