Assay Details
| Gene Symbol: RBL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101718.5 | NP_564058.1 | 3 - 4 | 980 | 131 | |
| GenBank mRNA | AF370504.1 | - | 3 - 4 | 784 | 131 | |
| AY081685.1 | - | 3 - 4 | 615 | 131 | ||
| AY086276.1 | - | 3 - 4 | 633 | 131 | ||
More Information
Set Membership: |
|

RBL12
Genomic Map