Assay Details
| Gene Symbol: AGL65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035989.1 | NP_001031066.1 | 1 - 2 | 119 | 68 | |
| NM_001332366.1 | NP_001321504.1 | 2 - 3 | 699 | 68 | ||
| NM_001332367.1 | NP_001321506.1 | 2 - 3 | 691 | 68 | ||
| NM_101733.6 | NP_173310.2 | 2 - 3 | 596 | 68 | ||
| GenBank mRNA | AY143172.1 | - | 1 - 2 | 184 | 68 | |
More Information
Set Membership: |
|

AGL65
Genomic Map