Assay Details
| Gene Symbol: AGL65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035989.1 | NP_001031066.1 | 7 - 8 | 433 | 125 | |
| NM_001332366.1 | NP_001321504.1 | 7 - 8 | 1112 | 125 | ||
| NM_001332367.1 | NP_001321506.1 | 8 - 9 | 1005 | 125 | ||
| NM_001332368.1 | NP_001321505.1 | 3 - 4 | 202 | 125 | ||
| NM_001332369.1 | NP_001321507.1 | 2 - 3 | 229 | 125 | ||
| NM_101733.6 | NP_173310.2 | 8 - 9 | 910 | 125 | ||
| GenBank mRNA | AY143172.1 | - | 7 - 8 | 498 | 125 | |
More Information
Set Membership: |
|

AGL65
Genomic Map