Assay Details
| Gene Symbol: AGL65 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035989.1 | NP_001031066.1 | 9 - 10 | 628 | 111 | |
| NM_001332366.1 | NP_001321504.1 | 9 - 10 | 1307 | 111 | ||
| NM_001332367.1 | NP_001321506.1 | 10 - 11 | 1200 | 111 | ||
| NM_001332368.1 | NP_001321505.1 | 5 - 6 | 397 | 111 | ||
| NM_001332369.1 | NP_001321507.1 | 4 - 5 | 424 | 111 | ||
| NM_101733.6 | NP_173310.2 | 10 - 11 | 1105 | 111 | ||
| GenBank mRNA | AY143172.1 | - | 9 - 10 | 693 | 111 | |
More Information
Set Membership: |
|

AGL65
Genomic Map