Assay Details
| Gene Symbol: AT1G18830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332373.1 | NP_001320897.1 | 20 - 21 | 2873 | 72 | |
| NM_001332374.1 | NP_001320898.1 | 20 - 21 | 2838 | 72 | ||
| NM_101740.3 | NP_173317.2 | 20 - 21 | 2815 | 72 | ||
More Information
Set Membership: |
|

AT1G18830
Genomic Map