Assay Details
| Gene Symbol: AT1G18960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101753.3 | NP_173330.1 | 1 - 2 | 220 | 68 | |
| GenBank mRNA | AB493466.1 | - | 1 - 2 | 118 | 68 | |
More Information
Set Membership: |
|

AT1G18960
Genomic Map