Assay Details
| Gene Symbol: TTN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198102.1 | NP_001185031.1 | 6 - 7 | 601 | 104 | |
| NM_101766.3 | NP_173342.2 | 5 - 6 | 470 | 104 | ||
| GenBank mRNA | BT010836.1 | - | 5 - 6 | 398 | 104 | |
| BT011312.1 | - | 4 - 5 | 306 | 104 | ||
More Information
Set Membership: |
|

TTN10
Genomic Map