Assay Details
| Gene Symbol: DA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332389.1 | NP_001320535.1 | 11 - 12 | 1733 | 83 | |
| NM_101785.4 | NP_173361.1 | 10 - 11 | 1926 | 83 | ||
More Information
Set Membership: |
|

DA1
Genomic Map