Assay Details
| Gene Symbol: DA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332389.1 | NP_001320535.1 | 4 - 5 | 738 | 126 | |
| NM_101785.4 | NP_173361.1 | 3 - 4 | 931 | 126 | ||
More Information
Set Membership: |
|

DA1
Genomic Map