Assay Details
| Gene Symbol: GAMMA CA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198110.1 | NP_001185039.1 | 3 - 4 | 560 | 82 | |
| NM_101815.4 | NP_564091.1 | 3 - 4 | 637 | 82 | ||
| GenBank mRNA | AK176887.1 | - | 3 - 4 | 443 | 82 | |
| AK229264.1 | - | 3 - 4 | 434 | 82 | ||
More Information
Set Membership: |
|

GAMMA CA1
Genomic Map