Assay Details
| Gene Symbol: AT1G19740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101830.2 | NP_173404.1 | 1 | 141 | 74 | |
| GenBank mRNA | AY128391.1 | - | 1 | 141 | 74 | |
| BT000129.1 | - | 1 | 67 | 74 | ||
More Information
Set Membership: |
|

AT1G19740
Genomic Map