Assay Details
| Gene Symbol: MP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332415.1 | NP_001321214.1 | 4 - 5 | 503 | 100 | |
| NM_101840.3 | NP_173414.1 | 3 - 4 | 1037 | 100 | ||
| GenBank mRNA | AF334716.1 | - | 3 - 4 | 353 | 100 | |
| BT002050.1 | - | 3 - 4 | 827 | 100 | ||
| BT008805.1 | - | 3 - 4 | 353 | 100 | ||
More Information
Set Membership: |
|

MP
Genomic Map