Assay Details
| Gene Symbol: SYTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332435.1 | NP_001319049.1 | 6 - 7 | 593 | 108 | |
| NM_001332436.1 | NP_001322430.1 | 6 - 7 | 595 | 108 | ||
| NM_001332437.1 | NP_001322431.1 | 6 - 7 | 588 | 108 | ||
| NM_001332438.1 | NP_001322429.1 | 6 - 7 | 593 | 108 | ||
| NM_101862.5 | NP_173436.4 | 5 - 6 | 612 | 108 | ||
| GenBank mRNA | FM213366.1 | - | 5 - 6 | 607 | 108 | |
More Information
Set Membership: |
|

SYTB
Genomic Map