Assay Details
| Gene Symbol: AT1G20640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332454.1 | NP_001319052.1 | 4 - 5 | 1334 | 66 | |
| NM_101915.3 | NP_173488.1 | 3 - 4 | 1267 | 66 | ||
| NM_202145.1 | NP_973874.1 | 4 - 5 | 1074 | 66 | ||
| GenBank mRNA | AY091015.1 | - | 3 - 4 | 1132 | 66 | |
| AY117237.1 | - | 2 - 3 | 831 | 66 | ||
More Information
Set Membership: |
|

AT1G20640
Genomic Map