Assay Details
| Gene Symbol: HMGB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035997.1 | NP_001031074.1 | 4 - 5 | 397 | 77 | |
| NM_001084100.1 | NP_001077569.1 | 4 - 5 | 397 | 77 | ||
| NM_101920.3 | NP_564123.1 | 4 - 5 | 425 | 77 | ||
| GenBank mRNA | AB493468.1 | - | 3 - 4 | 211 | 77 | |
| AF370248.1 | - | 4 - 5 | 394 | 77 | ||
| AK118543.1 | - | 4 - 5 | 309 | 77 | ||
| AY063064.1 | - | 3 - 4 | 211 | 77 | ||
| AY084731.1 | - | 4 - 5 | 77 | |||
More Information
Set Membership: |
|

HMGB2
Genomic Map