Assay Details
| Gene Symbol: AT1G20750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101927.4 | NP_173498.3 | 1 - 2 | 336 | 120 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G20750| Gene Symbol: AT1G20750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101927.4 | NP_173498.3 | 1 - 2 | 336 | 120 | |
Set Membership: |
|