Assay Details
| Gene Symbol: AT1G20760 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101928.7 | NP_173499.1 | 6 - 7 | 1984 | 96 | |
| GenBank mRNA | AK226951.1 | - | 6 - 7 | 1891 | 96 | |
More Information
Set Membership: |
|

AT1G20760
Genomic Map