Assay Details
| Gene Symbol: AT1G21070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101961.4 | NP_564133.1 | 2 - 3 | 734 | 116 | |
| GenBank mRNA | BT024726.1 | - | 2 - 3 | 317 | 116 | |
| KP872774.1 | - | 2 - 3 | 317 | 116 | ||
More Information
Set Membership: |
|

AT1G21070
Genomic Map