Assay Details
| Gene Symbol: AT1G21160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101970.4 | NP_173540.2 | 10 - 11 | 3676 | 119 | |
| GenBank mRNA | AB493471.1 | - | 9 - 10 | 2959 | 119 | |
More Information
Set Membership: |
|

AT1G21160
Genomic Map