Assay Details
| Gene Symbol: AT1G21160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101970.4 | NP_173540.2 | 4 - 5 | 2691 | 138 | |
| GenBank mRNA | AB493471.1 | - | 3 - 4 | 1974 | 138 | |
More Information
Set Membership: |
|

AT1G21160
Genomic Map