Assay Details
| Gene Symbol: AT1G21160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101970.4 | NP_173540.2 | 8 - 9 | 3054 | 108 | |
| GenBank mRNA | AB493471.1 | - | 7 - 8 | 2337 | 108 | |
More Information
Set Membership: |
|

AT1G21160
Genomic Map