Assay Details
| Gene Symbol: AT1G21380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101990.4 | NP_564138.1 | 2 - 3 | 448 | 81 | |
| GenBank mRNA | AY034974.1 | - | 2 - 3 | 357 | 81 | |
| AY062980.1 | - | 1 - 2 | 116 | 81 | ||
More Information
Set Membership: |
|

AT1G21380
Genomic Map