Assay Details
| Gene Symbol: AT1G21380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101990.4 | NP_564138.1 | 4 - 5 | 622 | 121 | |
| GenBank mRNA | AY034974.1 | - | 4 - 5 | 531 | 121 | |
| AY062980.1 | - | 3 - 4 | 290 | 121 | ||
More Information
Set Membership: |
|

AT1G21380
Genomic Map