Assay Details
| Gene Symbol: AT1G21380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101990.4 | NP_564138.1 | 5 - 6 | 733 | 74 | |
| GenBank mRNA | AY034974.1 | - | 5 - 6 | 642 | 74 | |
| AY062980.1 | - | 4 - 5 | 401 | 74 | ||
More Information
Set Membership: |
|

AT1G21380
Genomic Map