Assay Details
| Gene Symbol: AT1G21380 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101990.4 | NP_564138.1 | 9 - 10 | 1243 | 102 | |
| GenBank mRNA | AY034974.1 | - | 9 - 10 | 1152 | 102 | |
| AY062980.1 | - | 8 - 9 | 911 | 102 | ||
More Information
Set Membership: |
|

AT1G21380
Genomic Map