Assay Details
| Gene Symbol: EMB1144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036080.2 | NP_001031157.1 | 6 - 7 | 738 | 70 | |
| NM_001036081.2 | NP_001031158.1 | 6 - 7 | 738 | 70 | ||
| NM_103779.5 | NP_564534.1 | 6 - 7 | 802 | 70 | ||
| GenBank mRNA | AK227099.1 | - | 6 - 7 | 672 | 70 | |
| AY057519.1 | - | 6 - 7 | 726 | 70 | ||
| BT015785.1 | - | 6 - 7 | 604 | 70 | ||
More Information
Set Membership: |
|

EMB1144
Genomic Map