Assay Details
| Gene Symbol: FNR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126017.5 | NP_201420.1 | 2 - 3 | 523 | 83 | |
| GenBank mRNA | AY072112.1 | - | 2 - 3 | 414 | 83 | |
| AY096665.1 | - | 2 - 3 | 336 | 83 | ||
More Information
Set Membership: |
|

FNR1
Genomic Map