Assay Details
| Gene Symbol: FNR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085336.1 | NP_001078805.1 | 6 - 7 | 897 | 106 | |
| NM_126017.5 | NP_201420.1 | 6 - 7 | 902 | 106 | ||
| GenBank mRNA | AY072112.1 | - | 6 - 7 | 793 | 106 | |
| AY096665.1 | - | 6 - 7 | 715 | 106 | ||
More Information
Set Membership: |
|

FNR1
Genomic Map