Assay Details
| Gene Symbol: AT5G66390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126037.5 | NP_201440.1 | 2 - 3 | 699 | 94 | |
| GenBank mRNA | AK227717.1 | - | 2 - 3 | 484 | 94 | |
| BT008314.1 | - | 2 - 3 | 429 | 94 | ||
More Information
Set Membership: |
|

AT5G66390
Genomic Map