Assay Details
| Gene Symbol: AT5G66390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126037.5 | NP_201440.1 | 3 - 4 | 860 | 66 | |
| GenBank mRNA | AK227717.1 | - | 3 - 4 | 645 | 66 | |
| BT008314.1 | - | 3 - 4 | 590 | 66 | ||
More Information
Set Membership: |
|

AT5G66390
Genomic Map