Assay Details
| Gene Symbol: DAR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203708.1 | NP_001190637.1 | 3 - 4 | 704 | 94 | |
| NM_001345787.1 | NP_001331414.1 | 2 - 3 | 769 | 94 | ||
| NM_126059.5 | NP_201462.2 | 3 - 4 | 744 | 94 | ||
More Information
Set Membership: |
|

DAR7
Genomic Map