Assay Details
| Gene Symbol: DAR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345788.1 | NP_001330142.1 | 1 - 2 | 214 | 169 | |
| NM_001345789.1 | NP_001330144.1 | 1 - 2 | 52 | 169 | ||
| NM_126060.1 | NP_201463.1 | 1 - 2 | 40 | 113 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001345788.1, NM_001345789.1 transcripts.
Set Membership: |
|

DAR6
Genomic Map