Assay Details
| Gene Symbol: DAR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345788.1 | NP_001330142.1 | 3 - 4 | 1076 | 83 | |
| NM_001345789.1 | NP_001330144.1 | 3 - 4 | 914 | 83 | ||
| NM_001345790.1 | NP_001330143.1 | 3 - 4 | 1042 | 83 | ||
| NM_126060.1 | NP_201463.1 | 3 - 4 | 846 | 83 | ||
More Information
Set Membership: |
|

DAR6
Genomic Map