Assay Details
| Gene Symbol: DAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203709.1 | NP_001190638.1 | 2 - 3 | 553 | 118 | |
| NM_001345792.1 | NP_001331698.1 | 2 - 3 | 586 | 118 | ||
| NM_001345795.1 | NP_001331697.1 | 1 - 2 | 352 | 118 | ||
| NM_126062.2 | NP_201465.2 | 2 - 3 | 308 | 118 | ||
More Information
Set Membership: |
|

DAR3
Genomic Map