Assay Details
| Gene Symbol: DAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203709.1 | NP_001190638.1 | 4 - 5 | 726 | 82 | |
| NM_001345791.1 | NP_001331695.1 | 3 - 4 | 856 | 82 | ||
| NM_001345792.1 | NP_001331698.1 | 4 - 5 | 759 | 82 | ||
| NM_001345793.1 | NP_001331699.1 | 4 - 5 | 732 | 82 | ||
| NM_001345794.1 | NP_001331696.1 | 4 - 5 | 729 | 82 | ||
| NM_001345795.1 | NP_001331697.1 | 3 - 4 | 525 | 82 | ||
| NM_126062.2 | NP_201465.2 | 4 - 5 | 481 | 82 | ||
More Information
Set Membership: |
|

DAR3
Genomic Map