Assay Details
| Gene Symbol: DAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203709.1 | NP_001190638.1 | 7 - 8 | 1128 | 130 | |
| NM_001345791.1 | NP_001331695.1 | 6 - 7 | 1258 | 130 | ||
| NM_001345792.1 | NP_001331698.1 | 7 - 8 | 1161 | 130 | ||
| NM_001345793.1 | NP_001331699.1 | 7 - 8 | 1134 | 130 | ||
| NM_001345794.1 | NP_001331696.1 | 7 - 8 | 1131 | 130 | ||
| NM_001345795.1 | NP_001331697.1 | 6 - 7 | 927 | 130 | ||
| NM_126062.2 | NP_201465.2 | 7 - 8 | 883 | 130 | ||
More Information
Set Membership: |
|

DAR3
Genomic Map