Assay Details
| Gene Symbol: HB53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126068.3 | NP_201471.1 | 1 - 2 | 585 | 94 | |
| GenBank mRNA | AY683477.1 | - | 1 - 2 | 540 | 94 | |
| BT024847.1 | - | 1 - 2 | 454 | 94 | ||
More Information
Set Membership: |
|

HB53
Genomic Map