Assay Details
| Gene Symbol: CHR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126073.3 | NP_201476.1 | 13 - 14 | 2021 | 68 | |
| GenBank mRNA | AY099638.1 | - | 13 - 14 | 1967 | 68 | |
| BT002161.1 | - | 13 - 14 | 1868 | 68 | ||
More Information
Set Membership: |
|

CHR1
Genomic Map