Assay Details
| Gene Symbol: CHR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126073.3 | NP_201476.1 | 15 - 16 | 2221 | 81 | |
| GenBank mRNA | AY099638.1 | - | 15 - 16 | 2167 | 81 | |
| BT002161.1 | - | 15 - 16 | 2068 | 81 | ||
More Information
Set Membership: |
|

CHR1
Genomic Map