Assay Details
| Gene Symbol: SDH1-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126074.3 | NP_201477.1 | 4 - 5 | 680 | 64 | |
| GenBank mRNA | AF367341.1 | - | 4 - 5 | 583 | 64 | |
| AF436833.1 | - | 4 - 5 | 629 | 64 | ||
| AJ001809.1 | - | 4 - 5 | 553 | 64 | ||
| AY045674.1 | - | 4 - 5 | 581 | 64 | ||
| AY124812.1 | - | 4 - 5 | 511 | 64 | ||
More Information
Set Membership: |
|

SDH1-1
Genomic Map